missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRIG2 Monoclonal Antibody (990310)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561070

Product Code. 30535397

  • 419.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRIG2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
O94898
Lrig2
Chinese hamster ovary cell line CHO-derived recombinant human LRIG-2, Gly41-Thr805.
100 μg
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunocytochemistry
990310
Unconjugated
Lrig2
4632419I10Rik; BB096938; Kiaa0806; leucine rich repeats and immunoglobulin like domains 2; leucine-rich repeats and immunoglobulin like domains 2; leucine-rich repeats and immunoglobulin-like domains 2; leucine-rich repeats and immunoglobulin-like domains protein 2; LIG2; LIG-2; Lrig2; UFS2
Mouse
Protein A/G
RUO
9860
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.