missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRP1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568421
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
This gene encodes a member of the low density lipoprotein (LDL) receptor family. These receptors play a wide variety of roles in normal cell function and development due to their interactions with multiple ligands. Disruption of this gene has been reported in several types of cancer. [provided by RefSeq, Jun 2016].
Specifications
| LRP1B | |
| Polyclonal | |
| Unconjugated | |
| LRP1B | |
| 9630004P12Rik; EC 1.1.1.94; EC 3.4.21.9; LDL receptor related protein 1B; low density lipoprotein receptor related protein-deleted in tumor; low density lipoprotein receptor-related protein 1B; low density lipoprotein-related protein 1B (deleted in tumors); low-density lipoprotein receptor-related protein 1B; Low-density lipoprotein receptor-related protein-deleted in tumor; LRP1B; LRP-1B; LRP-deleted in tumors; LRPDIT; LRP-DIT | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53353, 94217 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9JI18, Q9NZR2 | |
| LRP1B | |
| Synthetic peptide corresponding to residues near the C terminal of human LRP1B. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction