missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRP4 Recombinant Rabbit Monoclonal Antibody (HL2080)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550374
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
LRP4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
This gene encodes a member of the low-density lipoprotein receptor-related protein family. The encoded protein may be a regulator of Wnt signaling. Mutations in this gene are associated with Cenani-Lenz syndrome.
Spezifikation
| LRP4 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with no preservative | |
| O75096, Q8VI56, Q9QYP1 | |
| LRP4 | |
| Recombinant fragment of mouse LRP4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2080 | |
| Unconjugated | |
| LRP4 | |
| 6430526J12Rik; CLSS; CMS17; Corin; D230026E03; Kiaa0816; LDL receptor related protein 4; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; low-density lipoprotein receptor-related protein 4; LRP10; Lrp4; LRP-4; mdig; Megf7; multiple epidermal growth factor-like domains 7; SOST2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 228357, 4038, 83469 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur