Learn More
LRPPRC Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-21175-100UL
Description
The LRPPRC antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LRPPRC. This antibody recognizes human, mouse, rat antigen. The LRPPRC antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| LRPPRC | |
| 1000 μg/mL | |
| 130 kDa leucine rich protein, CLONE 23970, GP130, LRP 130, LRP130, LRPPRC, LSFC | |
| 100 μL | |
| 10128 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |