Learn More
LRPPRC Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-21175-20UL
 View more versions of this product
Product Code. 18084495
  • 145.00€ / 20µL
Description
The LRPPRC antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LRPPRC. This antibody recognizes human, mouse, rat antigen. The LRPPRC antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
LRPPRC
1000 μg/mL
130 kDa leucine rich protein, CLONE 23970, GP130, LRP 130, LRP130, LRPPRC, LSFC
20 μL
10128
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa