Learn More
LRRC6 Monoclonal Antibody (OTI2E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805971S
Description
LRRC6 Monoclonal antibody specifically detects LRRC6 in Human samples. It is validated for Western BlotSpecifications
| LRRC6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86X45 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23639 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2E4 | |
| Unconjugated | |
| LRRC6 | |
| CILD19; leucine rich repeat containing 6; leucine rich repeat containing 6 (testis); leucine-rich repeat-containing 6 (testis); leucine-rich repeat-containing protein 6; leucine-rich repeat-containing protein 6; protein tilB homolog; leucine-rich testis-specific protein; LRRC6; Lrtp; Mc2; Protein tilB homolog; testis-specific leucine-rich repeat protein; Tslrp; Unknown (protein for MGC:138048) | |
| Full length human recombinant protein of human LRRC6 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.