Learn More
LRRC8A/SWELL1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17155-1-AP-20UL
Description
The LRRC8A/SWELL1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human LRRC8A/SWELL1. This antibody recognizes human antigen. The LRRC8A/SWELL1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| LRRC8A/SWELL1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 56262 | |
| IgG |
| Immunohistochemistry, ELISA | |
| 133 μg/mL | |
| Rabbit | |
| Primary | |
| Human |