Learn More
LRRC8A/SWELL1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17155-1-AP-20UL
 View more versions of this product
Product Code. 17422323
  • 145.00€ / 20µL
Description
The LRRC8A/SWELL1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human LRRC8A/SWELL1. This antibody recognizes human antigen. The LRRC8A/SWELL1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
LRRC8A/SWELL1
Polyclonal
Unconjugated
20 μL
56262
IgG
Immunohistochemistry, ELISA
133 μg/mL