Learn More
LRRCC1 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | LRRCC1 |
|---|---|
| Concentration | 650 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18069875
|
Proteintech
26317-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18009455
|
Proteintech
26317-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The LRRCC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LRRCC1. This antibody recognizes mouse, human antigen. The LRRCC1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| LRRCC1 | |
| Immunohistochemistry, ELISA | |
| Rabbit | |
| 85444 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 650 μg/mL | |
| Liquid | |
| Mouse, Human | |
| Clerc, KIAA1764, LRRCC1, SAP2 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.