missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
165.00€ - 476.00€
Specifications
| Antigen | LRRK2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24312256), Immunohistochemistry-Frozen, Flow (Cell Surface) reported in scientific literature (PMID 20483355), Knockout Validated, KnockDown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry, Immunoassay |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18735703
|
Novus Biologicals
NB110-58771SS |
0.025 mL |
165.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18715623
|
Novus Biologicals
NB110-58771 |
0.1 mL |
476.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRRK2 Polyclonal specifically detects LRRK2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Knockout Validated, Knockdown Validated.Specifications
| LRRK2 | |
| Western Blot 1:1000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24312256), Immunohistochemistry-Frozen, Flow (Cell Surface) reported in scientific literature (PMID 20483355), Knockout Validated, KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.05% Sodium Azide | |
| augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 | |
| LRRK2 | |
| IgG | |
| Affinity Purified | |
| Human, rat, mouse and primate. Predicted to react with porcine based on 100% sequence homology. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry, Immunoassay | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Primate | |
| Q5S007 | |
| 120892 | |
| A synthetic peptide made to a C-terminal region, within residues 2500-2527 of the human LRRK2 protein, conjugated to KLH. [Swiss-Prot# Q5S007] | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 286 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title