missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRN2 Monoclonal Antibody (6A9)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521045
This item is not returnable.
View return policy
Description
Peptide Sequence: RVQETHPYHI LLSWVTPPNT VSTNLTWSSA SSLRGQGATA LARLPRGTHS YNITRLLQAT EYWACLQVAF ADAHTQLACV WARTKEATSC HRALGDRPG.
The protein encoded by this gene belongs to the leucine-rich repeat superfamily. This gene was found to be amplified and overexpressed in malignant gliomas. The encoded protein has homology with other proteins that function as cell-adhesion molecules or as signal transduction receptors and is a candidate for the target gene in the 1q32.1 amplicon in malignant gliomas. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.
Specifications
| LRRN2 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| O75325 | |
| LRRN2 | |
| LRRN2 (NP_006329, 532 a.a. ~ 630 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| 6A9 | |
| Unconjugated | |
| LRRN2 | |
| 5730406J09Rik; fibronectin type III, immunoglobulin and leucine rich repeat domain 7; FIGLER7; GAC1; Glioma amplified on chromosome 1 protein; leucine rich and ankyrin repeats 1; leucine rich repeat neuronal 2; leucine rich repeat neuronal 5; leucine rich repeat protein 2, neuronal; leucine-rich repeat neuronal protein 2; Leucine-rich repeat neuronal protein 5; LRANK1; LRRN2; LRRN5; NLRR-2; UNQ256/PRO293 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10446 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction