missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRRTM3 Monoclonal Antibody (568318)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561497

Product Code. 30536094

  • 429.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Recent evidence suggests that LRRTM3 promotes the processing of amyloid-precursor protein (APP) by the beta-secretase BACE, leading to the proteolytic production of the A-beta42 peptide that is the main component of amyloid plaques. Furthermore, LRRTM3 maps to a region of chromosome 10 linked to both late-onset Alzheimer's disease (AD) and elevated plasma A-beta42 levels, suggesting that LRRTM3 is a functional and positional candidate gene for AD.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRRTM3
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and no preservative
Q86VH5
LRRTM3
Mouse myeloma cell line NS0-derived recombinant human LRRTM3, Glu31-Lys419.
100 μg
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin)
568318
Unconjugated
LRRTM3
9630044H04Rik; leucine rich repeat transmembrane neuronal 3; leucine-rich repeat transmembrane neuronal protein 3; LRRTM3; UNQ803/PRO1693
Mouse
Protein A/G
RUO
347731
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.