Learn More
Invitrogen™ Lsm10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595808
Description
Immunogen sequence: MAVSHSVKER TISENSLIIL LQGLQGRVTT VDLRDESVAH GRIDNVDAFM NIRLAKVTYT DRWGHQVKLD DLFVTGRNVR YVHIPDDVNI TSTIEQQLQI IHRVRNFGGK GQGRWEFPPK NCK; Positive Samples: Rat spleen, Rat lung, Rat testis; Cellular Location: Nucleus.
LSM10 gene ontology annotations related to this gene include positive regulation of G1/S transition of mitotic cell cycle.
Specifications
| Lsm10 | |
| Polyclonal | |
| Unconjugated | |
| LSM10 | |
| LSM10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-123 of human LSM10 (NP_1162701). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 0.92 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Lsm10; LSM10, U7 small nuclear RNA associated; MST074; MSTP074; U7 snRNA-associated Sm-like protein LSm10; U7 snRNP-specific Sm-like protein LSM10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 366468 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.