Learn More
Grant Instruments™ ecocool™ Refrigerated/Heated Circulating Bath
Product Code. 15499214
Change view
Click to view available options
Temperature Range:
-20°C to +100°C
-25°C to +150°C
Capacity (Metric):
5 L
6 L
Model:
ecocool™ 100
ecocool™ 150
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range Capacity (Metric) Model Unit Size
15499214 -20°C to +100°C 5 L ecocool™ 100 Each
15419224 -25°C to +150°C 6 L ecocool™ 150 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15499214 Supplier Grant Instruments™ Supplier No. ECOCOOL 100R
  • ONLINE EXCLUSIVE PRICE
    4465.00€
    4162.45€ / Pack of 1
    Save 302.55€
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Saves up to 80% against standard compressor units. Grant Instruments™ ecocool™ Refrigerated/Heated Circulating Baths are supplied assembled as ready to use kits, complete with accessory hosing, clips and connectors as standard.
  • Eco mode optimises the cooling control needed, enabling rapid cool down when required.
  • True energy saving against competitor units that only switch the compressor on or off.
  • Intuitive user interface
  • No-spill valved front drain.
  • Built in Britain to the highest specifications, 5 day dispatch, 4 year industry leading warranty.
  • Modern, sleek, attractive design
  • Thermostat and chiller work in harmony, neither will operate alone, eliminating any danger of overheating or freezing.
  • Single front switch for user convenience.
  • No side vents - locate to suit the user, not the unit.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 5 L
Cooling Capacity 250W at 20°C, 230W at 10°C, 185W at 0°C, 115W at -10°C, 40W at -10°C
Temperature Range -20°C to +100°C
Display 4 digit LED
Model ecocool™ 100
Temperature Presets 3
Temperature Stability 0.05°C
Warranty 4 Years
Width (Metric) 245 mm
Width (Metric) Interior 154 mm
Height (Metric) 640 mm
Depth (Metric) Interior 118 mm
Shipping Weight (Metric) 29 kg
Voltage 120/230 V
Frequency 50/60 Hz