missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LTA4H Monoclonal Antibody (24GB5535)
GREENER_CHOICE

Mouse Monoclonal Antibody

204.00€ - 580.00€

Specifications

Antigen LTA4H
Clone 24GB5535
Content And Storage -20°C
Applications Flow Cytometry, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275649
GREENER_CHOICE
View Documents
Invitrogen™
MA557440
100 μL
580.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275482
View Documents
Invitrogen™
MA557441
20 μL
204.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LTA4H Monoclonal Antibody for Western Blot

Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
Specifications

Specifications

LTA4H
-20°C
Monoclonal
Liquid
IgG2b
Human, Rat
LTA4H
Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27
LTA4H
Primary
Affinity chromatography
24GB5535
Flow Cytometry, Western Blot
Unconjugated
Mouse
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P09960, P30349
299732, 4048
Recombinant protein of human LTA4H (aa 1-611).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.