missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lubricating Oil Standard 13: Ca @ 0.100; P @ 0.010; S @ 0.450; Zn @ 0.250 wt% in Lubricating Oil, VHG™
Description
Lubricating Oil Standard 13: Ca @ 0.100; P @ 0.010; S @ 0.450; Zn @ 0.250 wt% in Lubricating Oil
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Calcium/Zinc/Sulfur |
| CAS | 7723-14-0, 7440-70-2, 7440-66-6, 7704-34-9 |
| Methods | XRF |
| Analyte or Component Names | Phosphorus/Calcium/Zinc/Sulfur |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | 20 cSt Mineral Oil |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Phosphorus 0.01%/Calcium 0.1%/Zinc 0.2516%/Sulfur 0.4504% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?