missing translation for 'onlineSavingsMsg'
Learn More

Lubricating Oil Standard 2: Ca @ 0.600; P @ 0.005; S @ 0.050; Zn @ 0.080 wt% in Lubricating Oil, VHG™

Product Code. 17903976
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17903976 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17903976 Supplier VHG Labs Supplier No. VHGLOIL2100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Lubricating Oil Standard 2: Ca @ 0.600; P @ 0.005; S @ 0.050; Zn @ 0.080 wt% in Lubricating Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphorus/Sulfur/Zinc/Calcium
CAS 7723-14-0, 7704-34-9, 7440-66-6, 7440-70-2
Methods XRF
Analyte or Component Names Phosphorus/Sulfur/Zinc/Calcium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 20 cSt Mineral Oil
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Phosphorus 0.0051%/Sulfur 0.0502%/Zinc 0.0804%/Calcium 0.6%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.