Description
Requires
Cytiva HiTrap Columns, sold separately.
- HiTrap column inlets have 1/16 in. female threads, while the outlets have 1/16 in. male threads
Specifications
Specifications
| Product Type | Union, Bottom |
| Quantity | 2 Pack |
| Material | Polypropylene |
| Column Type | Column Accesory |
| For Use With (Application) | Connect a syringe to the bottom of a HiTrap column |