Learn More
Euro Protection™ Lumbar Belt with Adjustable Straps
Product Code. 12013596
Change view
Click to view available options
Length (Metric):
116 to 141 cm
80 to 95 cm
95 to 118 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
12013596 80 to 95 cm Each
12023596 95 to 118 cm Each
12033596 116 to 141 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12013596 Supplier Euro Protection™ Supplier No. 78101
  • ONLINE EXCLUSIVE PRICE
    33.60€
    16.74€ / Pack of 1
    Save 16.86€
    50% Off
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket
Euro Protection™ Lumbar Belt with Adjustable Straps uses variable pressure and adjustable elastic shoulder straps to provide lumbar support.
  • Variable pressure and fastening system
  • Adjustable shoulder straps
  • To be worn outside another garment
Specifications
Product Type Lumbar Support Belt
Length (Metric) 80 to 95 cm
Includes Shoulder Straps