missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lunapark Antibody (11-1-R), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-32543
This item is not returnable.
View return policy
Description
Lunapark Monoclonal antibody specifically detects Lunapark in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Lunapark | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| 2310011O18Rik, 4921514L11Rik, DKFZp686G1442, KIAA1715, LNP, Lunapark, MGC102862, protein lunapark, Ul, ulnaless | |
| Recombinant protein within mouse Lunapark aa 198-425/ 425. (Uniprot: Q9C0E8) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 11-1-R | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 80856 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction