missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13056-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes the only known protein inhibitor of zinc-dependent metallocarboxypeptidases.Specifications
| LXN | |
| Polyclonal | |
| Unconjugated | |
| LXN | |
| ECI, latexin, LXN, Protein MUM, TCI | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17035, 56925, 59073 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P70202, Q64361, Q9BS40 | |
| Lxn | |
| LXN Fusion Protein Ag3726 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction