Learn More
LXN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13056-1-AP-20UL
 View more versions of this product
Product Code. 16821084
  • 145.00€ / 20µL
Description
This gene encodes the only known protein inhibitor of zinc-dependent metallocarboxypeptidases.
Specifications
Specifications
LXN
Polyclonal
Unconjugated
LXN
ECI, latexin, LXN, Protein MUM, TCI
Rabbit
Antigen Affinity Chromatography
RUO
17035, 56925, 59073
-20°C
Liquid
Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P70202, Q64361, Q9BS40
Lxn
LXN Fusion Protein Ag3726
20 μL
Primary
Rat, Human, Mouse