missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLX1A Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06545MFV610
This item is not returnable.
View return policy
Description
SLX1A Polyclonal antibody specifically detects SLX1A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SLX1A | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| SLX1, SLX1 structure-specific endonuclease subunit homolog A (S. cerevisiae), SLX1B | |
| Produced in rabbits immunized with E. coli-derived Human SLX1A fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 548593 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction