missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXR alpha/NR1H3 Rabbit anti-Human, Mouse, Rat, Clone: 7Z4H10, Novus Biologicals™
Rabbit Monoclonal Antibody
201.00€ - 497.00€
Specifications
| Antigen | LXR alpha/NR1H3 |
|---|---|
| Clone | 7Z4H10 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18391702
|
Bio-Techne
NBP3-16304-20UL |
20 μg |
201.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18304445
|
Novus Biologicals
NBP3-16304-100UL |
100 μg |
497.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LXR alpha/NR1H3 Monoclonal antibody specifically detects LXR alpha/NR1H3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| LXR alpha/NR1H3 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human LXR alpha/NR1H3 (Q13133). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 7Z4H10 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 10062 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title