missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-108 Monoclonal Antibody (eBio13G3-19D (13G3-19D)), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen M050T03B08
This item is not returnable.
View return policy
Description
Ly108 is the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6). Mouse Ly108 is expressed on NK, B cells, and T cells. Activation of Ly108 receptor enhances NK cell activity and IFN-gamma expression.
Specifications
| Ly-108 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q9ET39 | |
| SLAMF6 | |
| Affinity chromatography | |
| RUO | |
| 30925 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| IgG2a κ |
| Flow Cytometry | |
| eBio13G3-19D (13G3-19D) | |
| NovaFluor Blue 660-120S | |
| SLAMF6 | |
| Activating NK receptor; CD352; KAL1; KAL1b; KALI; KALIb; Ly108; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTBA; NTB-A; NTBA receptor; RGD1561848; SF2000; SLAM family member 6; Slamf6; UNQ6123/PRO20080 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction