Learn More
Invitrogen™ Ly-49H Monoclonal Antibody (3D10), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1586.00€ - 3585.00€
Specifications
| Antigen | Ly-49H |
|---|---|
| Clone | 3D10 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry, Functional Assay, Immunoprecipitation |
| Classification | Monoclonal |
Description
The MAb 3D10 recognizes Ly-49H, a member of the C-type lectin Ly-49 multigene family of receptors found on natural killer cells. Ly-49H, like Ly-49D, lacks the characteristic ITIM (immunoreceptor tyrosine-based inhibitory motif) and contains an arginine in the transmembrane domain implying these molecules act as activation receptors. Ly-49H is expressed in a subset of NK cells but not NKT (NK1.1CD3+) cells. Natural killer cells can express several Ly-49 proteins. It has been shown that there is preferential staining of Ly-49H with Ly-49D (cat 12-5783). Expression has been confirmed on C57BL/6 and NWNA but not Balb/c or DBA/2 mice. Other mice strains have not been tested. No crossreactivity to Ly-49A, C, D or G2 has been observed. Addition of MAb 3D10 to cell cultures induces lysis of target cells.
Ly49H is a member of the C-type lectin Ly-49 multigene family of receptors found on natural killer cells. Ly49H, like Ly49D, lacks the characteristic ITAM (immunoreceptor tyrosine-based inhibitory motif) and contains an arginine in the transmembrane domain implying these molecules act as activation receptors. Ly49H is expressed in a subset of NK cells but not NKT (NK1.1/CD3+) cells. Natural killer cells can express several Ly49 proteins.Specifications
| Ly-49H | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 16639 | |
| Primary | |
| 4°C | |
| Klra8 |
| 3D10 | |
| Flow Cytometry, Functional Assay, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Mouse | |
| Q60682 | |
| IgG1 κ | |
| Affinity Chromatography | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.