Learn More
Ly-6G/Ly-6C Antibody (RB6-8C5), PE/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-00441PECY7
Product Code. 18058957
  • 480.00€ / 0.10mL
Description
Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ly-6G/Ly-6C
Monoclonal
PE-Cyanine7
PBS with 0.05% Sodium Azide
LY6G
Normal mouse bone marrow.
0.1 mL
Primary
Studies indicate that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also reacts with an additional subpopulation of Ly-6C high cells.
Store at 4°C in the dark. Do not freeze.
Flow Cytometry
RB6-8C5
Flow Cytometry
GR, Gr-1, Ly-6, Ly6G
Rat
Protein A or G purified
Dendritic Cell Markers, Inflammation, Myeloid Cell Markers, Myeloid derived Suppressor Cell
546644.0
Mouse
IgG2b κ