missing translation for 'onlineSavingsMsg'
Learn More

Ly-6G/Ly-6C Antibody (RB6-8C5), PE, Novus Biologicals™

Rat Monoclonal Antibody

Brand:  Novus Biologicals NBP2-00441PE

Product Code. 18095899

  • 444.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ly-6G/Ly-6C
Monoclonal
PE
PBS with 0.05% Sodium Azide
LY6G
Normal mouse bone marrow.
0.1 mL
Primary
Studies indicate that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also reacts with an additional subpopulation of Ly-6C high cells.
Store at 4°C. Do not freeze.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay
RB6-8C5
Flow Cytometry 1:10 - 1:1000
GR, Gr-1, Ly-6, Ly6G
Rat
Protein A or G purified
Dendritic Cell Markers, Inflammation, Myeloid Cell Markers, Myeloid derived Suppressor Cell
546644.0
Mouse
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.