Learn More
Ly-6G/Ly-6C Antibody (RB6-8C5), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-00441PCP
Product Code. 18077298
  • 457.00€ / 0.10mL
Description
Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ly-6G/Ly-6C
Monoclonal
PerCP
PBS with 0.05% Sodium Azide
LY6G
Normal mouse bone marrow.
0.1 mL
Dendritic Cell Markers, Inflammation, Myeloid Cell Markers, Myeloid derived Suppressor Cell
546644
Mouse
Purified
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
RB6-8C5
Flow Cytometry
GR, Gr-1, Ly-6, Ly6G
Rat
Protein A or G purified
RUO
Primary
Studies indicate that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also reacts with an additional subpopulation of Ly-6C high cells.
Store at 4C in the dark.
IgG2b κ