Learn More
Ly-6G Rat, anti-Mouse, Biotin, Clone: 1A8, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60031BT
Encompass_Preferred
 View more versions of this product
Product Code. 17188631
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • The 1A8 antibody reacts with Ly-6G, a 21 - 25 kDa GPI-anchored protein, which together with the structurally related Ly-6C protein comprises the granulocyte receptor-1 antigen (Gr-1)
  • Gr-1 is expressed on monocytes, neutrophils and subsets of macrophages, plasmacytoid dendritic cells, and T cells
  • Monocytes in the bone marrow transiently express Gr-1 during development and the expression level is strongly correlated with granulocyte differentiation and maturation
  • In the periphery, Gr-1 is found predominantly on neutrophils and is a useful marker for these cells
  • The 1A8 antibody binds specifically to Ly-6G, whereas another commonly used antibody, clone RB6-8C5, binds to both Ly-6G and Ly-6C
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Neutrophil Enrichment Kit (Catalog #19762)
Specifications
Specifications
Ly-6G
Monoclonal
Biotin
500 μg
Mouse
Granulocytes and Subsets, Myeloid Cells
Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Functional Assay
1A8
Rat
Primary
IgG2a κ