missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G Rat, Clone: RB6-8C5, enQuireBio™
Rat Monoclonal Antibody
131.25€ - 248.85€
Specifications
| Antigen | Ly-6G |
|---|---|
| Clone | RB6-8C5 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15992448
|
EnquireBio
QAB77-100ug |
100 μg |
131.25€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15902458
|
EnquireBio
QAB77-500ug |
500 μg |
248.85€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Ly-6G | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 17067 | |
| IgG2b κ | |
| Protein A/G Purified |
| RB6-8C5 | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| NP_001238984 | |
| Ly6c1 | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title