missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly6K Antibody (CL2433) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36763PE
This item is not returnable.
View return policy
Description
Ly6K Monoclonal antibody specifically detects Ly6K in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Ly6K | |
| Monoclonal | |
| PE | |
| cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K | |
| This antibody was developed using a recombinant protein derived from Q17RY6, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL2433 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 54742 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction