missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYK5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37972-20ul
This item is not returnable.
View return policy
Description
LYK5 Polyclonal antibody specifically detects LYK5 in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| LYK5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| LYK5FLJ90524, NY-BR-96, protein kinase LYK5, Serologically defined breast cancer antigen NY-BR-96, STE20-like pseudokinase, STE20-related adapter protein, STE20-related adaptor protein, STE20-related kinase adapter protein alpha, STE20-related kinase adaptor alpha, Stlk, STRAD alpha, STRADPMSE | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-210 of human LYK5 (NP_699166.2).,, Sequence:, FLPEGGCYELLTVIGKGFEDLMTVNLARYKPTGEYVTVRRINLEACSNEMVTFLQGELHVSKLFNHPNIVPYRATFIADNELWVVTSFMAYGSAKDLICTHFMDGMNELAIAYILQGVLKALDYIHHMGYVHRSVKASHILISVDGKVYLSGLRSNLSMISHGQRQRVVHDFPKYSVKVLPWLSPE | |
| 20 μL | |
| Signal Transduction | |
| 92335 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction