missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Lymphocyte Serum-free Medium, KBM 581

Product Code. 17842571 Shop All Corning Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17842571 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17842571 Supplier Corning™ Supplier No. 88581CM

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The use of T-lymphocytes in adaptive immunotherapy is emerging as an important tool in the treatment of cancer and other clinical applications.

Corning™'s Lymphocyte Serum Free Medium (88-581-CM) supports the proliferation and activation of lymphocytes, including tumor infiltrating lymphocytes (TIL), Cytokine-Induced Killer (CIK) cells and Natural Killer (NK) cells. There is technical data to support the use in CAR-T as well as several other important immunotherapy applications. The 88-581-CM medium is specialized for both activation and expansion of human CIK cells. This enhanced version contains more nutrients, shows improved proliferation capacity, and produces a higher ratio of CIK cells (CD3+ CD56+).
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Quantity 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.