Learn More
Abcam Lymphoprep
Product Code. 32113242 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32113242 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32113242 Supplier Abcam Supplier No. ab286892250mL
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This product is manufactured by BioVision, an Abcam company and was previously called M1274 LymphoprepTM.
This product is manufactured by BioVision, an Abcam company and was previously called M1274 LymphoprepTM.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 250 mL