missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lymphotoxin beta R/TNFRSF3 Antibody (MM0461-3T24) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
218.00€ - 554.00€
Specifications
| Antigen | Lymphotoxin beta R/TNFRSF3 |
|---|---|
| Clone | MM0461-3T24 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18049636
|
Novus Biologicals
NBP2-11947-0.025MG |
0.025 mg |
218.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18768753
|
Novus Biologicals
NBP2-11947 |
0.1 mg |
554.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lymphotoxin beta R/TNFRSF3 Monoclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| Lymphotoxin beta R/TNFRSF3 | |
| LYOPH | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cytokine Research | |
| P36941 | |
| 4055 | |
| IgG1 | |
| Protein G purified |
| MM0461-3T24 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
| LTBR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title