Learn More
LYZ Monoclonal Antibody (OTI1D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506192
Description
Lysozyme is synthesized predomitly in reactive histiocytes rather than in resting, unstimulated phagocytes.Specifications
| LYZ | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P61626 | |
| LYZ | |
| Full length human recombit protein of human LYZ produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1D8 | |
| Unconjugated | |
| LYZ | |
| LYZF1, LZM | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4069 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.