missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LZTR1 Recombinant Rabbit Monoclonal Antibody (1P0U1)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

176.00€ - 436.00€

Specifications

Antigen LZTR1
Clone 1P0U1
Concentration 1.66 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283414
View Documents
Invitrogen™
MA557965
20 μL
176.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30284043
GREENER_CHOICE
View Documents
Invitrogen™
MA557966
100 μL
436.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: GGAADNTLPN ELHCYDVDFQ TWEVVQPSSD SEVGGAEVPE RACASEEVPT LTYEERVGFK KSRDVFGLDF GTTSAKQPTQ PASELPSGRL FHAAAVISDA MYIFGGTVDN NIRSGEMYRF QFSCYPKCTL HEDYGRLWES RQFCDVEFVL GEKEECVQGH VAIVTARSRW LRRKITQARE RLAQKLEQEA APVPREAPGV AAGGARPPLL HVAIREAEAR PFEVLMQFLY TDKIKYPRKG HVEDVL.

LZTR1, a member of the BTB-kelch superfamily, was initially described as a putative transcriptional regulator based on weak homology to members of the basic leucine zipper-like family, the encoded protein subsequently has been shown to localize exclusively to the Golgi network where it may help stabilize the Golgi complex. Deletion of this gene may be associated with DiGeorge syndrome, a developmental field defect involving the third and fourth pharyngeal pouches, causing the absence of thymus and parathyroid glands, congenital cardiac abnormalities and facial dysmorphism. LZTR1 is tyrosine phosphorylated and subsequently degraded upon induction of apoptosis.
Specifications

Specifications

LZTR1
1.66 mg/mL
ELISA, Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Q9CQ33
66863
Recombinant fusion protein containing a sequence corresponding to amino acids 300-545 of human LZTR1 (NP_006758.2).
Antibody
1P0U1
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Mouse
LZTR1
1200003E21Rik; AI591627; AW550890; BTBD29; fj87c05; leucine zipper like transcription regulator 1; leucine-zipper-like transcription regulator 1; leucine-zipper-like transcriptional regulator 1; leucine-zipper-like transcriptional regulator, 1; Lztr1; LZTR-1; MGC21205; NS10; si:dkey-98f17.2; SWNTS2; Tcfl2; wu:fj87c05; zgc:158250
LZTR1
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.