Learn More
M-cadherin Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29928-1-AP-20UL
Description
The M-cadherin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human M-cadherin. This antibody recognizes human, rat antigen. The M-cadherin antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| M-cadherin | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| Cadherin 14, Cadherin 15, CDH14, CDH15, CDH3, CDHM, M cadherin, MCAD, M-cadherin, MRD3, Muscle cadherin | |
| 20 μL | |
| 1013 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |