missing translation for 'onlineSavingsMsg'
Learn More

m-Tolualdehyde, 97%, stab. with 0.1% hydroquinone

Product Code. 11468196
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468196 100 g 100g
11448196 5 g 5g
11458196 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468196 Supplier Thermo Scientific Alfa Aesar Supplier No. A15313.22
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 620-23-5 | C8H8O | 120.151 g/mol

m-Tolualdehyde is used as a fragrance agent in food and cosmetics. It is also used as a reagent in the preparation of a wide range of pharmaceutical compounds such as anti-inflammatory agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Tolualdehyde is used as a fragrance agent in food and cosmetics. It is also used as a reagent in the preparation of a wide range of pharmaceutical compounds such as anti-inflammatory agents.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-23-5
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00003374
InChI Key OVWYEQOVUDKZNU-UHFFFAOYSA-N
Synonym m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde
PubChem CID 12105
ChEBI CHEBI:28476
IUPAC Name 3-methylbenzaldehyde
SMILES CC1=CC=CC(=C1)C=O

Specifications

Density 1.02
Boiling Point 198°C to 200°C
Flash Point 78°C (172°F)
Odor Almond-like
Refractive Index 1.541
Quantity 100 g
Beilstein 741964
Sensitivity Air Sensitive
Solubility Information Not miscible in water.
Formula Weight 120.15
Percent Purity 97%
Chemical Name or Material m-Tolualdehyde, stabilized with 0.1% hydroquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.