missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAFB Monoclonal Antibody (OTI2A6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526421
This item is not returnable.
View return policy
Description
MAFB Monoclonal Antibody for Western Blot, IHC (P), Flow
The protein encoded by this gene is a basic leucine zipper transcription factor that plays an important role in the regulation of lineage-specific hematopoiesis. The encoded nuclear protein represses ETS1-mediated transcription of erythroid-specific genes in myeloid cells. This gene contains no introns.
Specifications
| MAFB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y5Q3 | |
| MAFB | |
| Human recombinant protein fragment corresponding to amino acids 32-320 of MAFB produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A6 | |
| Unconjugated | |
| MAFB | |
| b-maf; c-Maf; kr; Kreisler; Kreisler (maf-related) leucine zipper protein; Kreisler maf-related leucine zipper homolog; Kreisler maf-related leucine zipper homolog (b-maf); KRML; Krml1; LOW QUALITY PROTEIN: transcription factor MafB; maf; MAF bZIP transcription factor B; Maf1; MAFB; maf-B; MAFB/Kreisler basic region/leucine zipper transcription factor; MCTO; MGC43127; segmentation protein Kr; si:ch73-337g9.1; transcription factor Maf; Transcription factor Maf-1; transcription factor MafB; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog B; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog b (paralog b); v-maf musculoaponeurotic fibrosarcoma oncogene family, protein B; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein B (avian); V-maf musculoaponeurotic fibrosarcoma oncogene homolog B | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9935 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction