missing translation for 'onlineSavingsMsg'
Learn More

MAGEH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen MAGEH1
Concentration 0.31 mg/mL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16890134
View Documents
Proteintech
12424-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16880134
View Documents
Proteintech
12424-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Melanoma-associated antigen (MAGE) are completely silent in normal tissues, with the exception of male germ cells, and, for some of them, placenta. These antigens ought to be strictly tumor specific, expressed in tumor cells of various histological types. Because of their specific expression on tumor cells, these antigens are of particular interest for antitumor immunotherapy. Genes of the MAGE family direct the expression of tumor antigens that are recognized on a human melanoma by autologous cytolytic T lymphocytes. Though the function of MAGEH1 is not fully elucidated, it may play a role in embryonal development and tumor transformation or aspects of tumor progression.
Specifications

Specifications

MAGEH1
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
Unconjugated
Rabbit
Rat, Mouse, Human
Q5PPP4, Q9H213, Q9NWG9
28986, 367767, 75625
MAGEH1 Fusion Protein Ag3090
Primary
-20°C
MAGEH1
0.31 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Apoptosis related protein 1, APR 1, APR1, MAGE H1 antigen, MAGEH, MAGEH1, melanoma antigen family H, 1, Melanoma associated antigen H1, Restin
MAGEH1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.