Description
The Applied Biosystems MagMAX CORE disruption modules are a sample preparation system designed to complement the MagMAX CORE Nucleic Acid Purification Kit. This modular technology is optimized for the purification of nucleic acids from a wide range of sample matrices commonly submitted for veterinary diagnostic testing. The MagMAX CORE disruption modules aid in the processing of hard-to-lyse organisms like Mycobacterium avium paratuberculosis (MAP).
Key features
- Convenience—a complete solution using CORE chemistry
- Confidence—optimized extraction of nucleic acids from difficult-to-lyse samples
- Flexible—purchase only what you need to lyse MAP and other hard-to-lyse organisms; 100 reaction tube module for lower throughput or 500 reaction plate module for higher throughput
- Ergonomy—streamlined sample preparation protocol versus the previous mechanical lysis module
- Sustainability—reduced tube consumption with use of high-throughput plate module
There are two options of module depending on workflow and throughput needs:
- MagMAX CORE Disruption Tubes Module, 100 reactions, contains bead beating tubes prefilled with zirconia beads for effective disruption of individual samples
- MagMAX CORE Disruption Plates Module, 500 reactions, contains 96-deep-well plates prefilled with zirconia beads and foil seal adapted for effective disruption of a higher number of samples
Specifications
Spécification
| Format | Plate |
| Isolation Technology | Mechanical Disruption |
| Scale | Maxi |
| Quantity | 500 Reactions |