missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Viral/Pathogen Ultra Enzyme Mix

Product Code. 16396582
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
1000 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16396582 50 mL 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16396582 Supplier Applied Biosystems™ Supplier No. A42366
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This enzyme mix is included in the MagMAX Viral/Pathogen Nucleic Acid Ultra Isolation Kit.

This enzyme mix is included in the MagMAX Viral/Pathogen Nucleic Acid Ultra Isolation Kit (Cat. No. A42356). It is made available separately for applications that require more enzyme mix than is provided in the kit.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5° to -30°C).
Sample Type Liquid Samples (e.g. Serum), Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast
Efficiency High
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
Product Line MagMAX
Product Type Viral/Pathogen Ultra Enzyme Mix
Quantity 50 mL
Shipping Condition Dry Ice
Target Viral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids
Volume (Metric) 50 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.