missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Bromide Hexahydrate, TRC

Product Code. 30468152
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468152 10 g 10g
30477737 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30468152 Supplier Toronto Research Chemicals Supplier No. TRCM11037510G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium Bromide Hexahydrate
CAS 13446-53-2
Quantity 10 g
Molecular Formula Br2Mg.6[H2O]
Synonym Magnesium Bromide Hexahydrate, Magnesium Dibromide Hexahydrate
InChI Formula InChI=1S/2BrH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2
SMILES Br[Mg]Br
Molecular Weight (g/mol) 292.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.