missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Carbonate, TRC

Product Code. 17959354
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17959354 50 g 50g
17977714 10 g 10g
17967354 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17959354 Supplier Toronto Research Chemicals Supplier No. TRCM11032550G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-93-0
Molecular Formula C O3 . Mg
Molecular Weight (g/mol) 84.31
Synonym Magnesium carbonate, Magnesium carbonate light, Hydrated basic magnesium carbonate
IUPAC Name magnesium;carbonate
SMILES [Mg+2].[O-]C(=O)[O-]

Specifications

Quantity 50 g
InChI Formula InChI=1S/CH2O3.Mg/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2
Formula Weight 83.9698
Chemical Name or Material Magnesium Carbonate (AS)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.