missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride, anhydrous, 99%

Product Code. 11371679
Change view
Click to view available options
Quantity:
1 kg
25 kg
Unit Size:
1kg
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371679 25 kg 25kg
11381679 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371679 Supplier Thermo Scientific Alfa Aesar Supplier No. 012315.C4
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

For Use As a Catalyst, Flocculating Agent, Various Magnesium Cements | CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Hygroscopic; May contain trace amount of Zr as black insoluble particles
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-30-3
Molecular Formula Cl2Mg
Molecular Weight (g/mol) 95.21
MDL Number MFCD00011106
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
PubChem CID 5360315
ChEBI CHEBI:6636
SMILES [Mg++].[Cl-].[Cl-]

Specifications

Melting Point 714°C
Color White to Yellow
Physical Form Crystals
Quantity 25 kg
Assay Percent Range 99%
Solubility Information Highly soluble in water with evolution of heat. Slightly soluble in acetone and pyridine.
IUPAC Name magnesium;dichloride
Formula Weight 95.22
Odor Odorless
Sensitivity Hygroscopic
Appearance White to Yellow Crystals
Density 2.32
Chemical Name or Material Magnesium Chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.