missing translation for 'onlineSavingsMsg'
Learn More

Magnesium fluoride hydrate, min 87% MgF2

Product Code. 11445223
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445223 2500 g 2500g
11435223 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445223 Supplier Thermo Scientific Alfa Aesar Supplier No. A12830.0E
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-40-6 | F2Mg | 62.30 g/mol

Magnesium fluoride is used in ceramics, optical lenses, glass manufacturing, and single crystals for polarizing prisms. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium fluoride is used in ceramics, optical lenses, glass manufacturing, and single crystals for polarizing prisms. It is also used in organic synthesis.

Solubility
Soluble in water (87 mg/L at 18°C ).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents and strong acids. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-40-6
Molecular Formula F2Mg
Molecular Weight (g/mol) 62.30
MDL Number MFCD00011108
InChI Key ORUIBWPALBXDOA-UHFFFAOYSA-L
IUPAC Name magnesium(2+) difluoride
SMILES [F-].[F-].[Mg++]

Specifications

Quantity 2500 g
Merck Index 14,5665
Solubility Information Soluble in water (87mg/L at 18°C ).
Formula Weight 62.30 (Anhydrous)
Odor Odorless
Assay ≥87% MgF2
Chemical Name or Material Magnesium fluoride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.