missing translation for 'onlineSavingsMsg'
Learn More

Magnesium potassium chloride

Product Code. p-7080858
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423244 100 g 100g
11433244 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423244 Supplier Thermo Scientific Alfa Aesar Supplier No. A10582.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15681-05-7 | Cl3KMg | 169.753 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15681-05-7
Molecular Formula Cl3KMg
MDL Number MFCD00156154
InChI Key JNZGLUUWTFPBKG-UHFFFAOYSA-K
Synonym magnesium potassium chloride, kcl mgcl2, acmc-1bu1g, magnesium 2+ ion potassium trichloride, magnesium 2+ potassium trichloride
PubChem CID 19903064

Specifications

Quantity 100 g
SMILES [Mg+2].[Cl-].[Cl-].[Cl-].[K+]
IUPAC Name magnesium;potassium;trichloride
Molecular Weight (g/mol) 169.753
Formula Weight 169.76
Chemical Name or Material Magnesium potassium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.