Learn More
Hanna Instruments™ Magnesium Reagent Water Testing Kit
Specifications
Specifications
| No. of Tests | 50 |
| Quantity | 50/Pk. |
| Range | 0 to 150mg/L |
| Test Method | Calmagite |
| Type | Magnesium Reagent |