missing translation for 'onlineSavingsMsg'
Learn More

Magnesium stearate

Product Code. 11423357
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423357 1000 g 1000g
11433357 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423357 Supplier Thermo Scientific Alfa Aesar Supplier No. A16244.0B
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 557-04-0 | C36H70MgO4 | 591.257 g/mol

Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ingredient in baby formulas. Further, it is used in the hydrogenation process. In addition to this, it is used to bind the sugar in hard candies like mints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ing
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-04-0
Molecular Formula C36H70MgO4
Molecular Weight (g/mol) 591.257
MDL Number MFCD00036391
InChI Key HQKMJHAJHXVSDF-UHFFFAOYSA-L
Synonym magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt
PubChem CID 11177
ChEBI CHEBI:9254
IUPAC Name magnesium;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2]

Specifications

Melting Point 148°C to 152°C
Odor Odorless
Quantity 1000 g
Merck Index 14,5690
Solubility Information Slightly soluble in benzene. Insoluble in water,alcohol and ether.
Formula Weight 591.27
Chemical Name or Material Magnesium stearate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.